Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEPAI, Mouse anti-Human, DyLight 488, Clone: PMEPA1/2696, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279891G
Description
TMEPAI Monoclonal antibody specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
TMEPAI | |
PMEPA1/2696 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
PMEPA1 | |
Recombinant full-length human TMEPAI protein | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Signal Transduction | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title