Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEPAI Rabbit anti-Human, Clone: PMEPA1/6421R, Novus Biologicals™
Rabbit Monoclonal Antibody
$301.00
Specifications
Antigen | TMEPAI |
---|---|
Clone | PMEPA1/6421R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB121966
|
Novus Biologicals
NBP30724620UG |
20 μg |
Each of 1 for $301.00
|
|
|||||
Description
TMEPAI Monoclonal specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
TMEPAI | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
10 mM PBS with 0.05% BSA | |
56937 | |
IgG κ | |
Protein A or G purified |
PMEPA1/6421R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein | |
Recombinant full-length human TMEPAI protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title