Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEPAI Rabbit anti-Human, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc TMEPA1101AP
Description
TMEPAI Polyclonal antibody specifically detects TMEPAI in Human samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
TMEPAI | |
Polyclonal | |
Unconjugated | |
PMEPA1 | |
2210418I02Rik; AW455466; Erg1.2; N4wbp4; Nedd4 WW binding protein 4; NEDD4 WW domain-binding protein 4; PMEPA1; Prostate transmembrane protein androgen induced 1; prostate transmembrane protein, androgen induced 1; protein TMEPAI; Solid tumor-associated 1 protein; STAG1; Tmepai; transmembrane prostate androgen-induced protein; transmembrane, prostate androgen induced RNA | |
Purified TMEPAI protein which was emulsified with adjuvants to acheive the desired antigenicity. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q969W9 | |
Rabbit | |
Affinity chromatography | |
RUO | |
56937 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction