Learn More
Aviva Systems Biology Corp TMF1 Antibody - N-terminal region (ARP58092_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58092_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMF1. Alternate names: ARA160, TMFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-62234