Learn More
TMIGD2 Mouse anti-Human, Alexa Fluor 647, Clone-953728, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB8316R025
Catalog No. FAB8316R025
Add to cart
This item is not returnable. View return policy
Description
Description
TMIGD2 Monoclonal antibody specifically detects TMIGD2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TMIGD2
953728
Flow Cytometry 5 uL/10^6 cells
Q96BF3
Mouse
IgG2b
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human TMIGD2/CD28H in direct ELISAs. Stains human TMIGD2/CD28H transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide.
CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2
HEK293 human embryonic kidney cell line-derived human TMIGD2/CD28H Met1-Gly150 Accession # Q96BF3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
126259
Human