Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMC4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548708

Catalog No. PA548708


Add to cart

Description

Description

Predicted to react with bovine and mouse based on sequence homology.

May mediate anti-apoptotic activity.
Specifications

Specifications

EMC4
Polyclonal
PBS with 0.09% sodium azide
Q5J8M3
ER membrane protein complex subunit 4, Cell proliferation-inducing gene 17 protein, Transmembrane protein 85, EMC4, TMEM85
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51234
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
EMC4
Liquid
EMC4
KLH conjugated synthetic peptide between 34-62 amino acids from the N-terminal region of human TMM85.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit