Learn More
TMOD2 293T Cell Overexpression Lysate 1 (Denatured), Abnova
Manufacturer:  Abnova Corporation H00029767T01
Catalog No. 89-002-145
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
NM_014548
6.8
Transient overexpression cell Lysate was tested with Anti-TMOD2 antibody (H00029767-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
TMOD2 293T Cell Transient Overexpression Lysate(Denatured)
38.72
pCMV-TMOD2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.