Learn More
TMP21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 151991AP
Catalog No. 50-173-0059
Add to cart
Description
Description
TMP21 is a ubiquitously expressed protein that is involved in vesicular targeting and protein transport. More recent experiments have shown that it is also a component in the presenilin complex and modulates the -gamma-secretase but not the epsilon-secretase cleavage activity of the amyloid precursor protein. The presenilin complex is composed of the proteins APH1, nicastrin, and PEN2 in addition to presenilin-1. Together, these proteins cleave the amyloid precursor protein at what is known as the -gamma- and epsilon-sites and can lead to the accumulation of the Abeta cleavage product that is associated with Alzheimer ft.s disease. Co-immunoprecipitation experiments using antibodies against these proteins also yielded TMP21 indicating that TMP21 may play a role in the regulation of this complex. Suppression of TMP21 expression by siRNA in transfected cells caused increased -gamma-secretase activity but not epsilon-secretase activity, and increased Abeta production, demonstrating that TMP21 can modulate -gamma-secretase activity.
Specifications
Specifications
TMP21
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P49755, Q63584, Q9D1D4
P24(DELTA), p24delta, S31I125, S31III125, TMED10, Tmp 21 I, TMP21, Transmembrane protein Tmp21
Rabbit
IgG
150 μL
-20°C
Primary
10972, 68581, 84599
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TMED10
Liquid
Tmed10
TMP21 Fusion Protein Ag7347
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat