Learn More
TMPH hydrochloride, Tocris
Neuronal nicotinic receptor antagonist
Manufacturer:  R&D Systems 2438/10
Quantity 10mg
 View more versions of this product
Catalog No. 24-381-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
305.88
TMPH hydrochloride
C16H31NO2·HCl
2,2,6,6-Tetramethylpiperidin-4-yl heptanoate
Specifications
849461-91-2
C16H31NO2·HCl
2,2,6,6-Tetramethylpiperidin-4-yl heptanoate
TMPH hydrochloride
>98%
305.88
10mg
Safety and Handling
Recommended Storage : Desiccate at Room Temperature