Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

TMPRSS11A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550810

Catalog No. PA550810



Description

Description

The antibody detects endogenous levels of total TMPRSS11A protein.

Probable serine protease which may play a role in cellular senescence. Overexpression inhibits cell growth and induce G1 cell cycle arrest.
Specifications

Specifications

TMPRSS11A
Polyclonal
Unconjugated
TMPRSS11A
Liquid
TMPRSS11A
Fusion protein corresponding to a region derived from internal residues of human transmembrane protease, serine 11A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q6ZMR5
ECRG1, HATL1, HESP
Rabbit
IgG
100 μL
-20°C
Primary
339967
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit