Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMPRSS11B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550811

Catalog No. PA550811


Add to cart

Description

Description

The antibody detects endogenous levels of total TMPRSS11B protein.

Probable serine protease.
Specifications

Specifications

TMPRSS11B
Polyclonal
Unconjugated
TMPRSS11B
Liquid
TMPRSS11B
Fusion protein corresponding to a region derived from internal residues of human transmembrane protease, serine 11B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.2 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q86T26
HATL5
Rabbit
IgG
100 μL
-20°C
Primary
132724
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit