Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMPRSS13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550808

Catalog No. PA550808


Add to cart

Description

Description

The antibody detects endogenous levels of total TMPRSS13 protein.

This gene encodes a member of the type II transmembrane serine protease family. Transmembrane serine proteases are regulated by protease inhibitors and known to function in development, homeostasis, infection, and tumorigenesis. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

TMPRSS13
Polyclonal
Unconjugated
TMPRSS13
Liquid
TMPRSS13
Fusion protein corresponding to residues near the C terminal of human transmembrane protease, serine 13
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.6 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9BYE2
MSP, TMPRSS11
Rabbit
IgG
100 μL
-20°C
Primary
84000
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit