Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMPRSS5 Mouse anti-Human, Clone: OTI4G4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504339

Catalog No. 50-167-5056


Add to cart

Description

Description

This gene encodes a protein that belongs to the serine protease family. Serine proteases are known to be involved in many physiological and pathological processes.
Specifications

Specifications

TMPRSS5
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9H3S3
SPINESIN
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80975
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4G4
Unconjugated
TMPRSS5
Liquid
TMPRSS5
Full length human recombit protein of human TMPRSS5 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit