Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMS-HT Kit TMS-HT (=HMDS and TMCS in Anhydrous Pyridine), 1mL&asterisk;8/Reaction vial, capacity 2mL&asterisk;8, TCI America™

Click to view available options
Quantity:
KIT

[Trimethylsilylating Reagent, for OH compounds]

MDL Number MFCD00148687
UN Number 2924
Physical Form Clear Liquid-Cloudy Liquid at 20°C
Color Yellow
Quantity KIT
Chemical Name or Material TMS-HT Kit TMS-HT (=HMDS and TMCS in Anhydrous Pyridine) 1 mL * 8 / Reaction vial, capacity 2 mL * 8 [Trimethylsilylating Reagent, for OH compounds]

TSCA : No

Recommended Storage : Refrigerator