Learn More
TMSB4X Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 198501AP
Catalog No. 50-173-2574
Add to cart
Description
Description
This gene encodes an actin sequestering protein which plays a role in regulation of actin polymerization. The protein is also involved in cell proliferation, migration, and differentiation. This gene escapes X inactivation and has a homolog on chromosome Y.
Specifications
Specifications
TMSB4X
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P20065, P62328, P62329
FX, PTMB4, Seraspenide, T beta 4, TB4X, THYB4, Thymosin beta 4, thymosin beta 4, X linked, TMSB4, TMSB4X
Rabbit
IgG
150 μL
-20°C
Primary
19241, 7114, 81814
Immunoprecipitation, Western Blot
Unconjugated
TMSB4X
Liquid
TMSB4X
TMSB4X Fusion Protein Ag13914
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat