Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMTC3 Rabbit anti-Human, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP305133100UL
Description
TMTC3 Polyclonal antibody specifically detects TMTC3 in Human samples. It is validated for Western BlotSpecifications
TMTC3 | |
Unconjugated | |
Purified | |
Rabbit | |
IgG | |
100 μL | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human |
Western Blot | |
PBS with 50% glycerol, pH7.3. | |
DKFZp686C0968, DKFZp686M1969, DKFZp686O22167, DKFZp686O2342, FLJ90492, SMILE, transmembrane and tetratricopeptide repeat containing 3, transmembrane and TPR repeat-containing protein 3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 615-914 of human TMTC3 (NP_861448.2). | |
Affinity purified | |
RUO | |
Primary | |
160418 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title