Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMTC3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen TMTC3
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096320
View Documents
Novus Biologicals
NBP305133100UL
100 μg Each for $320.71
Add to cart
 
NB096321
View Documents
Novus Biologicals
NBP30513320UL
20 μg Each for $138.37
Add to cart
 
Description

Description

TMTC3 Polyclonal antibody specifically detects TMTC3 in Human samples. It is validated for Western Blot
Specifications

Specifications

TMTC3
Unconjugated
Rabbit
Human
DKFZp686C0968, DKFZp686M1969, DKFZp686O22167, DKFZp686O2342, FLJ90492, SMILE, transmembrane and tetratricopeptide repeat containing 3, transmembrane and TPR repeat-containing protein 3
Recombinant fusion protein containing a sequence corresponding to amino acids 615-914 of human TMTC3 (NP_861448.2).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
160418
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit