Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TMTsixplex™ Isobaric Label Reagent Set

TMT™ Isobaric Mass Tagging Kits and Reagents enable multiplex quantitation of proteins extracted from cells and tissues using tandem mass spectrometry.

$4628.00

Specifications

For Use With (Application) Protein quantitation in Mass spectrometry
Storage Requirements Freezer (-5 to -30°C)
Product Type Isobaric Label Reagent Set
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PI90309 View Documents Thermo Scientific™
90309
Pack for $4,628.00
Add to cart
 
Description

Description

Features:
  • Powerful - Concurrent MS analysis of multiple samples increases sample throughput and enables relative quantitation of up to six different samples derived from cells, tissues or biological fluids
  • Consistent - Identical reagent structure and performance among TMTzero™, TMTduplex™, TMTsixplex™ and TMT10plex™ reagents allows efficient transition from method development to multiplex quantitation, enabling biomarker discovery research across platforms and datasets
  • Robust - Consistent chemistry allows efficient transition from method development to multiplex quantitation, enabling biomarker discovery research
  • Efficient - Amine-reactive NHS-ester activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
  • Compatible - Optimized for use with high resolution Thermo Scientific MS/MS platforms, such as Q Exactive, Orbitrap Elite™ and Orbitrap Fusion™ Tribrid™ instruments with data analysis fully supported by Proteome Discoverer™ 1.0 and above
Specifications

Specifications

Protein quantitation in Mass spectrometry
Freezer (-5 to -30°C)
Isobaric Label Reagent Set

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit