Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMX3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549227

Catalog No. PA549227

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Probable disulfide isomerase, which participates in the folding of proteins containing disulfide bonds. May act as a dithiol oxidase.
Specifications

Specifications

TMX3
Polyclonal
PBS with 0.09% sodium azide
Q96JJ7
Protein disulfide-isomerase TMX3, Thioredoxin domain-containing protein 10, Thioredoxin-related transmembrane protein 3, TMX3, KIAA1830, TXNDC10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54495
Western Blot
Unconjugated
TMX3
Liquid
TMX3
KLH conjugated synthetic peptide between 402-431 amino acids from the C-terminal region of human TMX3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit