Learn More
TMX4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 213481AP
Catalog No. 50-173-3380
Add to cart
Description
Description
This antibody is specific to TMX4.
This gene encodes a member of the disulfide isomerase (PDI) family of endoplasmic reticulum (ER) proteins that catalyze protein folding and thiol-disulfide interchange reactions. The encoded protein has an N-terminal ER-signal sequence, a catalytically active thioredoxin domain, one transmembrane domain and C-terminal ASP/GLU-rich calcium binding domain. Unlike most members of this gene family, it lacks a C-terminal ER-retention sequence. The encoded protein has been shown to have reductase activity in vitro.
Specifications
Specifications
TMX4
0.56 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8C0L0, Q9H1E5
KIAA1162, PDIA14, TMX4, TXNDC13, UNQ475/PRO938
Rabbit
IgG
150 μL
-20°C
Primary
296182, 52837, 56255
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TMX4
Liquid
Tmx4
TMX4 Fusion Protein Ag15696
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat