Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNF-alpha, Human anti-Human, APC, Clone: cA2 (Infliximab), Novus Biologicals
Human Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275898APC
Description
TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human samples. It is validated for Flow Cytometry.Specifications
TNF-alpha | |
cA2 (Infliximab) | |
Flow (Intracellular) | |
Conjugated, Purified | |
TNF | |
Purified recombinant human TNF (rTNF). | |
Protein A purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Flow Cytometry | |
APC | |
PBS with 0.05% Sodium Azide | |
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
Human | |
IgG1 Kappa | |
0.1 mL | |
Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
Primary | |
Binds to soluble and transmembrane forms of TNF-alpha and neutralizes its biological activity. Does not bind to TNF-beta. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title