Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF-alpha Human anti-Human, DyLight™ 550, Clone: cA2 (Infliximab), Novus Biologicals™

Human Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275898R

Catalog No. NBP275898R


Add to cart

Description

Description

TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human samples. It is validated for ELISA.
Specifications

Specifications

TNF-alpha
cA2 (Infliximab)
ELISA
Conjugated, Purified
TNF
Purified recombinant human TNF (rTNF).
Protein A purified
Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Primary
7124.0
Human
ELISA
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Human
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Binds to soluble and transmembrane forms of TNF-alpha and neutralizes its biological activity. Does not bind to TNF-beta.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit