Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF-alpha Human anti-Human, FITC, Clone: CDP 571 (Humicade), Novus Biologicals™

Human Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275908F

Catalog No. NBP275908F


Add to cart

Description

Description

TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human samples. It is validated for ELISA, Immunohistochemistry.
Specifications

Specifications

TNF-alpha
CDP 571 (Humicade)
ELISA, Immunohistochemistry
Conjugated, Purified
TNF
Human TNF-alpha
Protein A purified
Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Primary
7124.0
Human
ELISA, Immunohistochemistry
FITC
PBS with 0.05% Sodium Azide
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Human
IgG4 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
The antibody binds to human TNF-alpha with a Kd of 0.087 nM.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit