Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF-alpha Mouse anti-Human, Alexa Fluor™ 350, Clone: 6N2E12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP227351AF350

Catalog No. NP22731A30


Add to cart

Description

Description

TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

TNF-alpha
6N2E12
Western Blot, Flow Cytometry
Conjugated, Purified
TNF
Full-length recombinant human TNF alpha was used as the immunogen for this antibody.
Protein G purified
Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Primary
7124.0
Flow Cytometry, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit