Learn More
TNF Mouse anti-Human, Clone: MAb1, Abnova™
Mouse monoclonal antibody raised against human TNF.
Manufacturer:  Abnova Corporation MAB12620
Catalog No. 89-935-023
Add to cart
Description
Description
This gene encodes a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophages. It can bind to, and thus functions through its receptors TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. This cytokine is involved in the regulation of a wide spectrum of biological processes including cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer. Knockout studies in mice also suggested the neuroprotective function of this cytokine. [provided by RefSeq]
Specifications
Specifications
TNF
MAb1
Mouse monoclonal antibody raised against human TNF.
In aqueous buffer (Stabilizer, 0.09% sodium azide).
DIF/TNF-alpha/TNFA/TNFSF2
Mouse
IgG1, kappa
50 ug
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (PFA fixed)
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) Sandwich ELISA The optimal working dilution should be determined by the end user.
Liquid
TNF
Human TNF
Affinity purification
RUO
Primary
7124