Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF Mouse anti-Human, Rat, Clone: TNF706, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14620

Catalog No. 89-034-703


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human TNF.

  • High specificity
  • High reactivity
Specifications

Specifications

TNF
TNF706
Mouse monoclonal antibody raised against partial recombinant human TNF.
In 10 mM PBS.
Liquid
TNF
Recombinant protein corresponding to N-terminus of human TNF.
Protein A/G purification
RUO
Primary
7124
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
P01375
DIF|TNF-alpha|TNFA|TNFSF2
Mouse
IgM, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)