Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF Rat anti-Mouse, BV421, Clone: MP6-XT22, BD Horizon™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 566287

Catalog No. BDB566287


Add to cart

Description

Description

TNF Monoclonal antibody specifically detects TNF in Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications

Specifications

TNF
MP6-XT22
Brilliant Violet 421
Purified
Rat
Rat IgG1
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions
Monoclonal
Flow Cytometry, Flow Cytometry (Intracellular Staining)
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Tnf; Tnfa; TNF alpha; TNF-a; Tnfsf1a; Tnfsf2; TNFSF2; Cachectin; DIF
Recombinant Mouse TNF
25μg
Cell Biology
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Reseach Use Only.