Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TNF RI/TNFRSF1A Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793105PLATES

 View more versions of this product

Catalog No. NB009660


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TNF RI/TNFRSF1A.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
7.81 pg/mL
7132
RUO
TNF RI/TNFRSF1A
Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Death Receptor Signaling Pathway, Phospho Specific, Signal Transduction
Human
TNFRSF1A
1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to a working concentration of 1.0 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 500 pg/mL is recommended
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TNF RI/TNFRSF1A.
Sandwich ELISA
  • Standard: Each vial contains 60 ng of recombinant anti-TNF RI/TNFRSF1A.
  • Capture Antibody: 0.5 mg/mL of mouse anti-TNF RI/TNFRSF1A monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL mouse anti-TNF RI/TNFRSF1A monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Storage is content dependent.
    Mouse
    5 Plates
    7.81-500 pg/mL
    CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit