Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TNF RI/TNFRSF1A Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6251SP
This item is not returnable.
View return policy
Description
TNF RI/TNFRSF1A Monoclonal specifically detects TNF RI/TNFRSF1A in Human samples. It is validated for Western Blot.Specifications
TNF RI/TNFRSF1A | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNFRI, TNF-RI, TNF-R-I, TNFR-I, TNFRSF1A, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor | |
E. coli-derived recombinant human TNF RI/TNFRSF1A Arg235-Arg455 Accession # P19438 | |
25 μg | |
Primary | |
Detects human TNF RI/TNFRSF1A in direct ELISAs. | |
Human | |
Purified |
Western Blot | |
925224 | |
Western Blot 2 ug/mL | |
P19438 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
7132 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction