Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF RI/TNFRSF1A Mouse anti-Human, Clone: 925224, R&D Systems™

Mouse MonoclonalAntibody

$116.82 - $341.00

Specifications

Antigen TNF RI/TNFRSF1A
Clone 925224
Dilution Western Blot 2 ug/mL
Applications Western Blot
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
MAB6251100 View Documents R&D Systems
MAB6251100
100 ug Unconjugated Each for $341.00
Add to cart
 
MAB6251SP View Documents R&D Systems
MAB6251SP
25 ug Unconjugated Each for $116.82
Add to cart
 
Description

Description

TNF RI/TNFRSF1A Monoclonal antibody specifically detects TNF RI/TNFRSF1A in Human samples. It is validated for Western Blot.
Specifications

Specifications

TNF RI/TNFRSF1A
Western Blot 2 ug/mL
Purified
Human
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
E. coli-derived recombinant human TNF RI/TNFRSF1A Arg235-Arg455 Accession # P19438
Monoclonal
Protein A or G purified from ascites
Detects human TNF RI/TNFRSF1A in direct ELISAs.
925224
Western Blot
Mouse
RUO
7132
IgG2a
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit