Learn More
Aviva Systems Biology Corp TNFAIP8L1 Antibody - middle region (ARP52919_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52919_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNFAIP8L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TNFAIP8L1. Alternate names: MGC17791, TIPE1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Catalog No. 50-014-60870