Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFAIP8L3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP310186100UL

 View more versions of this product

Catalog No. NB125273


Add to Cart

Description

Description

TNFAIP8L3 Polyclonal specifically detects TNFAIP8L3 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

TNFAIP8L3
Polyclonal
Western Blot 1.0 ug/ml
TNF Alpha-Induced Protein 8-Like Protein 3, TNFAIP8-Like Protein 3, Tumor Necrosis Factor Alpha-Induced Protein 8-Like Protein 3, Tumor Necrosis Factor, Alpha-Induced Protein 8-Like 3, Tumor Necrosis Factor, Alpha-Induced Protein 8-Like Protein 3
The immunogen is a synthetic peptide directed towards the middle region of mouse TNFAIP8L3 (NP_001028707.1). Peptide sequence NVLSKLLHECKDLVHELVQRHLTPRTHGRINHVFNHFADVEFLSTLYGPH
100 μg
Primary
Mouse
Purified
Western Blot
Unconjugated
PBS buffer, 2% sucrose
Rabbit
Affinity purified
RUO
388121
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit