Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNFRH3/TNFRSF26 Rat anti-Mouse, Alexa Fluor 405, Clone: 509027, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB5330V100UG
This item is not returnable.
View return policy
Description
TNFRH3/TNFRSF26 Monoclonal specifically detects TNFRH3/TNFRSF26 in Mouse samples. It is validated for Flow Cytometry.Specifications
TNFRH3/TNFRSF26 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P83626 | |
Rat | |
100 μg | |
Primary | |
Detects mouse TNFRH3/TNFRSF26 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Fas, rmTNF RI, and rmTNF RII is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
509027 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Tnfrsf26 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse TNFRH3/TNFRSF26 Ser20-Lys162 Accession # P83626 | |
RUO | |
8718 | |
Mouse | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)