Learn More
TNFRSF10A Mouse anti-Human, Clone: 32A242, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human TNFRSF10A.
Manufacturer:  Abnova Corporation MAB12969
Catalog No. 89-935-026
Add to cart
Description
Description
The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is activated by tumor necrosis factor-related apoptosis inducing ligand (TNFSF10/TRAIL), and thus transduces cell death signal and induces cell apoptosis. Studies with FADD-deficient mice suggested that FADD, a death domain containing adaptor protein, is required for the apoptosis mediated by this protein. [provided by RefSeq]
Specifications
Specifications
TNFRSF10A
32A242
Mouse monoclonal antibody raised against synthetic peptide of human TNFRSF10A.
In PBS (0.2% gelatin, 0.05% sodium azide).
APO2/CD261/DR4/MGC9365/TRAILR-1/TRAILR1
Mouse
IgG1
50 ug
Store at 4°C. Do not freeze.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
TNFRSF10A
A synthetic peptide corresponding to amino acids 2-21 of human TNFRSF10A.
Protein G purification
RUO
Primary
8797