Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR4 Mouse anti-Human, FITC, Clone: DR-4-02, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen A15746

Catalog No. 50-113-7745


Add to cart

Description

Description

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors, TNFR1 and Fas. A novel member in the TNF family was recently identified and designated TRAIL (for TNF-related apoptosis-inducing ligand) and Apo-2L (for Apo-2 ligand). TRAIL is a type II membrane protein and expressed in a variety of human tissues. Two novel death domain containing receptors DR4 and DR5 have been identified as the receptor for TRAIL. Like TNF and Fas ligand, TRAIL induces apoptosis and NF-βB activation in many tissues and cells.
Specifications

Specifications

DR4
Monoclonal
0.1 mg/mL
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
O00220
APO2, CD261, DR4, MGC9365, TRAILR-1, TRAILR1
Mouse
IgG1
100 μg
4° C
Primary
8797
Flow Cytometry
DR-4-02
FITC
TNFRSF10A
Liquid
TNFRSF10A
Human CD261
Purified
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit