Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF11B Mouse anti-Human, Clone: 98A1071, Abnova™

Mouse monoclonal antibody raised against human TNFRSF11B.

Manufacturer:  Abnova Corporation MAB12971

Catalog No. 89-935-029


Add to cart

Description

Description

The protein encoded by this gene is a member of the TNF-receptor superfamily. This protein is an osteoblast-secreted decoy receptor that functions as a negative regulator of bone resorption. This protein specifically binds to its ligand, osteoprotegerin ligand, both of which are key extracellular regulators of osteoclast development. Studies of the mouse counterpart also suggest that this protein and its ligand play a role in lymph-node organogenesis and vascular calcification. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined. [provided by RefSeq]

Sequence: TQETFPPKYLHYDEETSH
Specifications

Specifications

TNFRSF11B
98A1071
Mouse monoclonal antibody raised against human TNFRSF11B.
In PBS (0.05% BSA, 0.05% sodium azide).
MGC29565/OCIF/OPG/TR1
Mouse
IgG
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
TNFRSF11B
A synthetic peptide corresponding to amino acids 20-37 of human TNFRSF11B.
Protein G purification
RUO
Primary
4982
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit