Learn More
TNFRSF13C Mouse anti-Human, Clone: 11C1, Abnova™
Mouse monoclonal antibody raised against human TNFRSF13C.
Manufacturer:  Abnova Corporation MAB12949
Catalog No. 89-935-030
Add to cart
Description
Description
B cell-activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. Overexpression of Baff in mice results in mature B-cell hyperplasia and symptoms of systemic lupus erythematosus (SLE). Also, some SLE patients have increased levels of BAFF in serum. Therefore, it has been proposed that abnormally high levels of BAFF may contribute to the pathogenesis of autoimmune diseases by enhancing the survival of autoreactive B cells. The protein encoded by this gene is a receptor for BAFF and is a type III transmembrane protein containing a single extracellular cysteine-rich domain. It is thought that this receptor is the principal receptor required for BAFF-mediated mature B-cell survival. [provided by RefSeq]
Specifications
Specifications
TNFRSF13C
11C1
Mouse monoclonal antibody raised against human TNFRSF13C.
In PBS, pH 7.2 (0.09% sodium azide).
BAFF-R/BAFFR/CD268/MGC138235
Mouse
IgG1
50 ug
Store at 4°C. Do not freeze.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
TNFRSF13C
Human TNFRSF13C.
Affinity purification
RUO
Primary
115650