Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ TNFRSF18 (Human) ELISA Kit
Sandwich enzyme-linked immunosorbent assay
Supplier: Abnova™ KA5688
Description
TNFRSF18 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of human TNFRSF18 in cell culture supernates, cell lysates, serum and plasma (heparin, EDTA).Specifications
Human | |
TNFRSF18 | |
8784 | |
RUO |
Quantitative | |
AITR, GITR, GITR-D | |
1 Kit | |
Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction