Learn More
TNFRSF18 Mouse anti-Human, Clone: OTI4H5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806389
Catalog No. 50-166-7817
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Glucocorticoid-induced tumor necrosis factor receptor family-related gene (GITR) is a member of the tumor necrosis factor receptor (TNFR) family that is expressed at low levels on unstimulated T cells, B cells, and macrophages. Upon activation, CD4+ and CD8+ T cells up-regulate GITR expression, whereas immunoregulatory T cells constitutively express high levels of GITR.
Specifications
Specifications
TNFRSF18
Monoclonal
Unconjugated
TNFRSF18
Lyophilized
TNFRSF18
Human recombit protein fragment corresponding to amino acids 26-162 of human TNFRSF18 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI4H5
PBS with 8% trehalose and no preservative; pH 7.3
Q9Y5U5
AITR, CD357, GITR, GITR-D
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8784