Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNFRSF18 Mouse anti-Human, Clone: UMAB203, liquid, UltraMAB™

Mouse Monoclonal Antibody
Supplier: OriGene UM870095

Description
Glucocorticoid-induced tumor necrosis factor receptor family-related gene (GITR) is a member of the tumor necrosis factor receptor (TNFR) family that is expressed at low levels on unstimulated T cells, B cells, and macrophages. Upon activation, CD4+ and CD8+ T cells up-regulate GITR expression, whereas immunoregulatory T cells constitutively express high levels of GITR.Specifications
TNFRSF18 | |
Monoclonal | |
Unconjugated | |
TNFRSF18 | |
AITR, CD357, GITR, GITR-D | |
Mouse | |
Affinity Chromatography | |
RUO | |
8784 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
UMAB203 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9Y5U5 | |
TNFRSF18 | |
Human recombit protein fragment corresponding to amino acids 26-162 of human TNFRSF18 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title