Learn More
TNFRSF19 Mouse anti-Human, Clone: OTI1B7, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810325
Catalog No. 50-167-0013
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TROY can mediate activation of c-Jun and NF-kappa-B. May promote caspase-independent cell death. Isoform 2 and isoform 3 may act as decoy receptors.
Specifications
Specifications
TNFRSF19
Monoclonal
Unconjugated
TNFRSF19
Lyophilized
TNFRSF19
Human recombit protein fragment corresponding to amino acids 30-170 of human TNFRSF19 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1B7
PBS with 8% trehalose and no preservative; pH 7.3
Q9NS68
TAJ, TAJ-alpha, TRADE, TROY
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
55504