Learn More
Abnova™ TNFRSF21 Recombinant Protein
Human TNFRSF21 full-length ORF ( AAH05192.1, 1 a.a. - 124 a.a.) recombinant protein with GST-tag at N-terminal
$503.00 - $775.00
Specifications
Accession Number | AAH05192.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 27242 |
Molecular Weight (g/mol) | 39.38 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-012-571
|
Abnova™
H00027242P0110 |
10 ug |
Each for $503.00
|
|
|||||
89-012-572
|
Abnova™
H00027242P0125 |
25 ug |
Each for $775.00
|
|
|||||
Description
The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor has been shown to activate NF-kappaB and MAPK8/JNK, and induce cell apoptosis. Through its death domain, this receptor interacts with TRADD protein, which is known to serve as an adaptor that mediates signal transduction of TNF-receptors. Knockout studies in mice suggested that this gene plays a role in T-helper cell activation, and may be involved in inflammation and immune regulation. (provided by RefSeq)
- Molecular weight: 39.38
- Preparation method: in vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8 in the elution buffer
- Quality Control Testing:12.5% SDS-PAGE stained with Coomassie Blue
Best use within three months from the date of receipt of this protein
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH05192.1 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
39.38 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TNFRSF21 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
27242 | |
TNFRSF21 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQFNFELSFKYVLYSSYSWLKLDHTIADCMVFTWTPCRMLDYLYSSYANMLWAGEMKSSSHQDLLFKWLDNWATKELELHLLGFELFWNTLLHFGKSKSSASGALSIENLPSFALKDVLFFIYT | |
BM-018/DR6/MGC31965 | |
TNFRSF21 | |
Wheat Germ (in vitro) | |
GST |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.