Learn More
Sino Biological™ TNFRSF4 Recombinant Human Protein, hIgG1-Fc.His Tag Non-distribution product as customer accommodation.
Expressed from DNA sequence encoding extracellular domain (Met 1-Ala 216) of human TNFRSF4 precursor fused to C-terminal polyhistidine-tagged Fc region of human IgG1
Manufacturer:  Sino Biological™ 10481H03H50
 View more versions of this product
Catalog No. 10-481-H03H50
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description
Expressed from DNA sequence encoding extracellular domain (Met 1-Ala 216) of human TNFRSF4 precursor fused to C-terminal polyhistidine-tagged Fc region of human IgG1
  • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Description & Specifications
Specifications
Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
22163
>85% by SDS-PAGE
Human
<1 EU/μg
TNFRSF4
Cytokines and Receptors, CD Proteins
TNFRSF4 Recombinant Human Protein
Lyophilized
48.2kd
50μg
Truncated, Recombinant
Tumor Necrosis Factor Receptor Superfamily, Member 4
Assay Standard
Mammalian
hIgG1-Fc.His Tag