Sino Biological™ TNFRSF4 Recombinant Human Protein, hIgG1-Fc.His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding extracellular domain (Met 1-Ala 216) of human TNFRSF4 precursor fused to C-terminal polyhistidine-tagged Fc region of human IgG1

Manufacturer: Sino Biological™ 10481H03H25

 View more versions of this product

Catalog No. 10-481-H03H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Structural Form Truncated, Recombinant
    Molecular Weight 48.2kd
    Purity >85% by SDS-PAGE
    Quantity 5 x 5μg
    Source Human
    Endotoxin Concentration <1 EU/μg
    Common Name Tumor Necrosis Factor Receptor Superfamily, Member 4
    Gene Symbol TNFRSF4
    Protein Tag hIgG1-Fc.His Tag
    Validated Application Assay Standard
    Protein Family Cytokines and Receptors, CD Proteins
    Expression System Mammalian
    Product Type TNFRSF4 Recombinant Human Protein
    For Use With (Application) Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 22163

    Expressed from DNA sequence encoding extracellular domain (Met 1-Ala 216) of human TNFRSF4 precursor fused to C-terminal polyhistidine-tagged Fc region of human IgG1

    • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit