Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNFRSF8, anti-Human, Clone: Ki-3, Abnova™
Monoclonal Antibody
Supplier: Abnova Corporation RAB00140
Specifications
TNFRSF8 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.2 (0.02% Proclin 300). | |
CD30/D1S166E/KI-1 | |
Original antibody is raised against native purified human TNFRSF8. | |
RUO | |
943 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
Ki-3 | |
ELISA (0.1 ug/mL) Flow Cytometry (1 ug/mL) Immunofluorescence Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
tumor necrosis factor receptor superfamily, member 8 | |
TNFRSF8 | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction