Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF8, anti-Human, Clone: Ki-4, Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00128

Catalog No. 89-032-471


Add to cart

Specifications

Specifications

TNFRSF8
Ki-4
ELISA (0.1 ug/mL) Flow Cytometry (1 ug/mL) Immunofluorescence Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
tumor necrosis factor receptor superfamily, member 8
CD30|D1S166E|KI-1
Original antibody is raised against native purified human TNFRSF8.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
943
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Liquid
TNFRSF8
IgG, kappa
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit