Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF8 Mouse anti-Human, Clone: Ber-H2, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14195

Catalog No. 89-034-296


Add to cart

Description

Description

Mouse monoclonal antibody raised against native human TNFRSF8.

  • High specificity
  • High reactivity
Specifications

Specifications

TNFRSF8
Ber-H2
Mouse monoclonal antibody raised against native human TNFRSF8.
In tissue culture supernatant (0.05% sodium azide).
Liquid
TNFRSF8
Co cell line established from a patient with Hodgkin's disease of T-cell lineage.
500 uL
Store at 4°C.
Monoclonal
The epitope of this antibody is located between amino acids 112 - 412 of human TNFRSF8.
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (5-10 uL/million cells in 0.1 mL) Immunofluorescence (1:50-1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:100) The optimal working dilution should be determined by the end user.
P28908
CD30|D1S166E|KI-1
Mouse
IgG1, kappa
RUO
Primary
943
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)