Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF8 Mouse anti-Human, Clone: MEM-268, Abnova™

Mouse monoclonal antibody raised against human TNFRSF8.

Manufacturer:  Abnova Corporation MAB12937

Catalog No. 89-935-035


Add to cart

Description

Description

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is expressed by activated, but not by resting, T and B cells. TRAF2 and TRAF5 can interact with this receptor, and mediate the signal transduction that leads to the activation of NF-kappaB. This receptor is a positive regulator of apoptosis, and also has been shown to limit the proliferative potential of autoreactive CD8 effector T cells and protect the body against autoimmunity. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq]

Specifications

Specifications

TNFRSF8
MEM-268
Mouse monoclonal antibody raised against human TNFRSF8.
In PBS, pH 7.4 (0.09% sodium azide).
CD30/D1S166E/KI-1
Mouse
IgG
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
TNFRSF8
THP-1 cell line.
Protein A purification
RUO
Primary
943
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit