Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNFSF15 Mouse anti-Human, Clone: VEGI/1283, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14902
Description
Mouse monoclonal antibody raised against full length recombinant human TNFSF15.
- High specificity
- High reactivity
Specifications
TNFSF15 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
MGC129934/MGC129935/TL1/TL1A/VEGI/VEGI192A | |
Mouse | |
Protein A/G purification | |
RUO | |
9966 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
VEGI/1283 | |
Mouse monoclonal antibody raised against full length recombinant human TNFSF15. | |
In 10 mM PBS. | |
TNFSF15 | |
Recombinant protein corresponding to full-length human TNFSF15. | |
100 μg | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction