Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFSF15 Rat anti-Mouse, Clone: Tan 2-2, MilliporeSigma™

Rat Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen TNFSF15
Clone Tan 2-2
Concentration 1.0 mg/mL
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Neutralization, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABC1628100
View Documents
MilliporeSigma
MABC1628100UG
100 μg
Each for $373.00
Add to cart
 
MABC162825
View Documents
MilliporeSigma
MABC162825UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

TNFSF15 Monoclonal antibody specifically detects TNFSF15 in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry, Neutralizing, Western Blotting.
Specifications

Specifications

TNFSF15
1.0 mg/mL
Unconjugated
Rat
Mouse
Recombinant fragment corresponding to 176 amino acids from the extracellular domain of murine TNFSF15 TL1A protein with a human IgG1 Fc domain and an additional hinge-like region at the C-terminus.
Monoclonal
Protein G purified
Tan 2-2
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Neutralization, Western Blot
Purified
RUO
Purified rat monoclonal antibody IgG2a in PBS without preservatives.
IgG2aκ
Primary
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit